Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45367

Sigma-Aldrich

N-(2,3-Epoxypropyl)phthalimide

≥95.0% (GC)

Synonym(s):

N-Glycidylphthalimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO3
CAS Number:
Molecular Weight:
203.19
Beilstein:
171277
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95.0% (GC)

form

solid

mp

92-98 °C (lit.)

SMILES string

O=C1N(CC2CO2)C(=O)c3ccccc13

InChI

1S/C11H9NO3/c13-10-8-3-1-2-4-9(8)11(14)12(10)5-7-6-15-7/h1-4,7H,5-6H2

InChI key

DUILGEYLVHGSEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service