Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

453242

Sigma-Aldrich

2-(Chloromethyl)phenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2C6H4NCO
CAS Number:
Molecular Weight:
167.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.567 (lit.)

bp

45-47 °C/0.2 mmHg (lit.)

density

1.241 g/mL at 25 °C (lit.)

SMILES string

ClCc1ccccc1N=C=O

InChI

1S/C8H6ClNO/c9-5-7-3-1-2-4-8(7)10-6-11/h1-4H,5H2

InChI key

XTYOTPBIDRGAML-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmad Basheer et al.
The Journal of organic chemistry, 71(26), 9743-9750 (2006-12-16)
The reaction of omega-chloroalkyl isocyanates Cl(CH2)nNCO (n = 2 (2), 3 (4)) and isothiocyanate Cl(CH2)2NCS (3) with active methylene compounds CH2YY' 1 in the presence of Et3N or Na give 2-YY'-methylene-1,3-oxazolidines, (E,Z)-1,3-thiazolidines, and 1,3-oxazines from 2, 3, and 4, respectively.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service