Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

453153

Sigma-Aldrich

Tetrakis(diethylamido)zirconium(IV)

electronic grade, ≥99.99% trace metals basis

Synonym(s):

Tetrakis(diethylamido)zirconium, Tetrakis(diethylamino)zirconium, Zirconium tetrakis(diethylamide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C2H5)2N]4Zr
CAS Number:
Molecular Weight:
379.74
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

Assay

≥99.99% trace metals basis

form

liquid

reaction suitability

core: zirconium

refractive index

n20/D 1.51 (lit.)

bp

128 °C/0.05 mmHg (lit.)

density

1.026 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCN(CC)[Zr](N(CC)CC)(N(CC)CC)N(CC)CC

InChI

1S/4C4H10N.Zr/c4*1-3-5-4-2;/h4*3-4H2,1-2H3;/q4*-1;+4

InChI key

GOVWJRDDHRBJRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 40 Zirconium

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service