Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

452831

Sigma-Aldrich

3-Bromo-1,1,1-trichloropropane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2CCl3
CAS Number:
Molecular Weight:
226.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.513 (lit.)

bp

113-114 °C (lit.)

density

1.762 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)(Cl)CCBr

InChI

1S/C3H4BrCl3/c4-2-1-3(5,6)7/h1-2H2

InChI key

PUNXOOKBNWKKKR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

676. Fluoro-olefins. Part II. Synthesis and reactions of some 3: 3: 3-trihalogenopropenes.
Haszeldine RN.
Journal of the Chemical Society, 3371-3378 (1953)
Negative ion formation in dissociative electron attachment to selected halogen derivatives of propane.
Barszczewska W, et al.
International Journal of Mass Spectrometry, 277(1), 103-106 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service