Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

451894

Sigma-Aldrich

Sodium mercury amalgam

beads, ≥99.9%, Na 10 %

Synonym(s):

Sodium amalgam

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥99.9%

form

beads

composition

Na, 10%

reaction suitability

reagent type: reductant

mp

223 °C

SMILES string

[Na].[Hg]

InChI

1S/Hg.Na

InChI key

MJGFBOZCAJSGQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Magnetic characteristics were studied of organometallic oxo-vanadium cluster compounds, such as [C5Me5V]4(μ-O)6, which is formed by reduction with sodium mercury amalgam.

Used for:
  • Ionic liquid based sodium ion conducting gel polymer electrolytes
  • Dehalogenation agent for the room-temperature synthesis of carbonaceous films
  • Plant-scaled method for preparation of deuterium-depleted water
  • Investigations of solvent effects on desulfonylation of polyethylene glycol-substituted aryl alkyl sulfones

Catalyst for synthesis of selectively benzylated Me α-D-glucopyranosides

Reagent for reduction of RDX for functionalization for forensic analysis

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - STOT RE 1 - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abernethy, C.D. et al.
Organometallics, 18, 870-870 (1999)
Yuesong Wang et al.
Analytical chemistry, 80(7), 2599-2605 (2008-02-26)
We report new assays of enzymes uroporphyrinogen decarboxylase (UROD) and coproporphyrinogen III oxidase (CPO) in the heme biosynthetic pathway. The assays were developed for use in clinical diagnostics of inherited disorders porphyria cutanea tarda and hereditary coproporphyria, respectively. Electrospray ionization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service