Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

451878

Sigma-Aldrich

4-Mercapto-1-butanol

95%

Synonym(s):

4-Hydroxy-1-mercaptobutane, 4-Hydroxybutanethiol, 4-Mercapto-1-butanol, 4-Mercaptobutan-1-ol, 4-Mercaptobutanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)4OH
CAS Number:
Molecular Weight:
106.19
Beilstein:
1731412
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.494 (lit.)

bp

102-103 °C/20 mmHg (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

OCCCCS

InChI

1S/C4H10OS/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

NEJMTSWXTZREOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to form a hydrophilic sparse ionic monolayer to protected metal nanoparticles to create Highly Concentrated Nano-Au and Nano-Ag "Inks" that can be Sintered to Near-Bulk Conductivity at 150 °C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gulcin Bolat et al.
Sensors (Basel, Switzerland), 13(7), 8079-8094 (2013-06-26)
Herein, we report a simple and inexpensive way for the fabrication of an ultramicroelectrode and present its characterization by electrochemical techniques. The fabrication of polyaniline UME involves only two steps: modification of a gold (Au) electrode by self assembled monolayers
Edyta Mikuła et al.
BMC veterinary research, 14(1), 328-328 (2018-11-08)
In this work, we report an electrochemical biosensor for the detection of anti-hemagglutinin antibodies against the swine virus H1N1 present in mice sera immunized with mixture of His6-H1 HA in monomeric and oligomeric form. The oriented immobilization of the recombinant
Bibin, A. T.; Sivaramakrishnan, S.; Chua, L.-L.; Ho, P. K. H.
Advances in Functional Materials, 20, 296-303 (2010)
Kamila Malecka et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-06)
The background: The monolayers self-assembled on the gold electrode incorporated transition metal complexes can act both as receptor ("host" molecules) immobilization sites, as well as transducer for interface recognitions of "guest" molecules present in the aqueous solutions. Their electrochemical parameters
Santiago Oviedo-Rouco et al.
Archives of biochemistry and biophysics, 680, 108243-108243 (2020-01-04)
Cytochrome c is a prototypical multifunctional protein that is implicated in a variety of processes that are essential both for sustaining and for terminating cellular life. Typically, alternative functions other than canonical electron transport in the respiratory chain are associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service