Skip to Content
MilliporeSigma
All Photos(1)

Documents

451630

Sigma-Aldrich

Gallium(III) fluoride

anhydrous, powder, ≥99.99% trace metals basis

Synonym(s):

Gallium fluoride, Gallium trifluoride, Trifluorogallium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaF3
CAS Number:
Molecular Weight:
126.72
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: gallium

density

4.47 g/mL at 25 °C (lit.)

SMILES string

F[Ga](F)F

InChI

1S/3FH.Ga/h3*1H;/q;;;+3/p-3

InChI key

WXXZSFJVAMRMPV-UHFFFAOYSA-K

Application

Employed in the preparation and study of optical fluorogallate glasses.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Park et al.
Biochimica et biophysica acta, 1430(1), 127-140 (1999-03-20)
Magnesium (Mg2+) is the physiological divalent cation stabilizing nucleotide or nucleotide analog in the active site of myosin subfragment 1 (S1). In the presence of fluoride, Mg2+ and MgADP form a complex that traps the active site of S1 and
Journal of Materials Research, 11, 2633-2633 (1996)
B Bureau et al.
Solid state nuclear magnetic resonance, 14(3-4), 181-190 (1999-09-28)
Owing to the implementation of acquisition techniques specific for nuclei with very large quadrupolar interaction (full shifted echo and variable offset cumulative spectra (VOCS)), NMR spectra of 69Ga and 71Ga are obtained in crystallised (PbGaF5, Pb3Ga2F12, Pb9Ga2F24 and CsZnGaF6) and
B Bureau et al.
Solid state nuclear magnetic resonance, 15(2), 129-138 (2000-02-12)
Owing to the implementation of acquisition techniques specific for nuclei with very large quadrupolar interaction (full shifted echo and variable offset cumulative spectra (VOCS)), NMR spectra of 69Ga and 71Ga are obtained in crystallised (PbGaF5, Pb3Ga2F12, Pb9Ga2F24 and CsZnGaF6) and
Tomonao Inobe et al.
Journal of molecular biology, 329(1), 121-134 (2003-05-14)
To understand the mechanism of a functionally important ATP-induced allosteric transition of GroEL, we have studied the effect of a series of metal fluoride-ADP complexes and vanadate-ADP on GroEL by kinetic fluorescence measurement of pyrene-labeled GroEL and by small-angle X-ray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service