Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

449911

Sigma-Aldrich

Magnesium iodide

AnhydroBeads, −10 mesh, 99.998% trace metals basis

Synonym(s):

Magnesium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgI2
CAS Number:
Molecular Weight:
278.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.998% trace metals basis

product line

AnhydroBeads

Quality Level

Assay

99.998% trace metals basis

impurities

≤25.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

637 °C (dec.) (lit.)

density

4.43 g/mL at 25 °C (lit.)

SMILES string

I[Mg]I

InChI

1S/2HI.Mg/h2*1H;/q;;+2/p-2

InChI key

BLQJIBCZHWBKSL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Anhydrous magnesium iodide, when reacted with cyclic sulfates of vic-diols, produces the corresponding olefins in excellent yields.[1]

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jang, D.O. Joo, Y.H.
Synthetic Communications, 28, 871-871 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service