All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
99%
refractive index
n20/D 1.398 (lit.)
bp
84 °C/745 mmHg (lit.)
density
0.983 g/mL at 25 °C (lit.)
SMILES string
CC1COCO1
InChI
1S/C4H8O2/c1-4-2-5-3-6-4/h4H,2-3H2,1H3
InChI key
SBUOHGKIOVRDKY-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
28.4 °F - closed cup
Flash Point(C)
-2 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Polymerizability of methyl substituted 1, 3-dioxolanes.
Okada M, et al.
Makromol. Chem., 176(4), 859-872 (1975)
Further studies on the relative ease of reductive cleavage of 1, 3-dioxolanes and 1, 3-dioxanes in ether solution by LiAlH4-AlCl3.
Diner UE and Brown RK.
Canadian Journal of Chemistry, 45(11), 1297-1299 (1967)
Acidolysis of poly (4-methyl-1, 3-dioxolane).
Diab MA, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 25(4), 945-954 (1987)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service