Skip to Content
MilliporeSigma
All Photos(1)

Documents

449377

Sigma-Aldrich

Di-2-thienyl ketone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6OS2
CAS Number:
Molecular Weight:
194.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

90-94 °C (lit.)

SMILES string

O=C(c1cccs1)c2cccs2

InChI

1S/C9H6OS2/c10-9(7-3-1-5-11-7)8-4-2-6-12-8/h1-6H

InChI key

GUTQMBQKTSGBPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed Bakir et al.
Talanta, 62(4), 781-789 (2008-10-31)
The compound di-2-thienyl ketone p-nitrophenylhydrazone (DSKNPH) melting point 168-170 degrees C was isolated in good yield from the reaction between di-2-thienyl ketone (DSK) and p-nitrophenylhydrazine in refluxing ethanol containing a few drop of concentrated HCl. Nuclear magnetic resonance studies on
Photophysical properties of some thienyl ketones: an experimental and theoretical study.
Becker RS, et al.
The Journal of Physical Chemistry, 99(5), 1410-1417 (1995)
Perthienyl analogues of chichibabin's hydrocarbon. A new stable, highly amphoteric multistage redox system with low oxidation potentials.
Kawase T, et al.
Tetrahedron Letters, 33(37), 5405-5408 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service