Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

448613

Sigma-Aldrich

Dichloro(2-chloroethyl)methylsilane

97%

Synonym(s):

(2-Chloroethyl)-dichloro-methylsilane, (2-Chloroethyl)methyl-dichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7Cl3Si
CAS Number:
Molecular Weight:
177.53
Beilstein:
1734992
EC Number:
MDL number:
UNSPSC Code:
12352000
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.458 (lit.)

density

1.241 g/mL at 20 °C (lit.)

SMILES string

C[Si](Cl)(Cl)CCCl

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service