Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

448540

Sigma-Aldrich

(S)-(−)-α-Methyl-4-pyridinemethanol

99%

Synonym(s):

(S)-(−)-1-(4-Pyridyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO
CAS Number:
Molecular Weight:
123.15
Beilstein:
1524527
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D −58°, c = 1 in chloroform

optical purity

ee: 98% (GLC)

mp

67-69 °C (lit.)

functional group

hydroxyl

SMILES string

C[C@H](O)c1ccncc1

InChI

1S/C7H9NO/c1-6(9)7-2-4-8-5-3-7/h2-6,9H,1H3/t6-/m0/s1

InChI key

HVOAMIOKNARIMR-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service