Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

448400

Sigma-Aldrich

2-Phenoxyethylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CH2NH2
CAS Number:
Molecular Weight:
137.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.536 (lit.)

bp

101-103 °C/4 mmHg (lit.)

density

1.048 g/mL at 25 °C (lit.)

SMILES string

NCCOc1ccccc1

InChI

1S/C8H11NO/c9-6-7-10-8-4-2-1-3-5-8/h1-5H,6-7,9H2

InChI key

IMLAIXAZMVDRGA-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stable gaseous aziridinium ions.
Van de Sande CC, et al.
Org. Mass Spectrom., 13(11), 666-670 (1978)
A R Katritzky et al.
The Journal of organic chemistry, 66(16), 5590-5594 (2001-08-04)
1-[2-Arylthio(oxy)ethyl]-5-benzotriazolyl-2-pyrrolidinones 6a-e, 12 and 3-benzotriazolyl-2-[2-arylthio(oxy)ethyl]-1-isoindolinones 9a-f, 14 are readily available from reactions of benzotriazole (4), 2-(arylsulfanyl)ethylamines 3, or 2-phenoxyethylamine (11) with 2,5-dimethoxy-2,5-dihydrofuran (5) or 2-formylbenzoic acid (8). Lewis acid mediated cyclizations of 6 and 9 produced novel 1,4-benzothiazepines 7a-e and
Synthesis and Characterization of Imino-Phenolate Titanium Complexes and Their Use in Homo-and Copolymerization of Ethylene.
da S Gomes F, et al.
Macromolecular Chemistry and Physics, 215(18), 1735-1743 (2014)
Dynamic nuclear polarization with polychlorotriphenylmethyl radicals: supramolecular polarization-transfer effects.
Cristina Gabellieri et al.
Angewandte Chemie (International ed. in English), 49(19), 3360-3362 (2010-04-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service