447625
Hexaphenyldigermanium(IV)
97%
Synonym(s):
Hexaphenyldigermane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C6H5)3GeGe(C6H5)3
CAS Number:
Molecular Weight:
607.90
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay
97%
reaction suitability
core: germanium
mp
340 °C (lit.)
SMILES string
c1ccc(cc1)[Ge](c2ccccc2)(c3ccccc3)[Ge](c4ccccc4)(c5ccccc5)c6ccccc6
InChI
1S/C36H30Ge2/c1-7-19-31(20-8-1)37(32-21-9-2-10-22-32,33-23-11-3-12-24-33)38(34-25-13-4-14-26-34,35-27-15-5-16-28-35)36-29-17-6-18-30-36/h1-30H
InChI key
MZZSGDPNYJAKCA-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service