Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

447145

Sigma-Aldrich

Borane dimethyl sulfide complex solution

5.0 M in diethyl ether

Synonym(s):

BMS, Trihydro[thiobis[methane]]boron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S · BH3
CAS Number:
Molecular Weight:
75.97
Beilstein:
3663489
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: reductant

concentration

5.0 M in diethyl ether

bp

~34 °C

density

0.789 g/mL at 25 °C

functional group

thioether

storage temp.

2-8°C

SMILES string

B.CSC

InChI

1S/C2H6S.BH3/c1-3-2;/h1-2H3;1H3

InChI key

RMHDLBZYPISZOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Borane dimethyl sulfide complex can be used in the preparation of:
  • An amine-based polymer for CO2 uptake.
  • Amine-modified electrospun ultrafine fibers used to remove methyl orange from water.
  • Thexylborane, a key intermediate for the synthesis of polyethylene derived star copolymers and terpolymers.

Reactant used as a regioselective reducing agent

Reactant involved in:
  • Hydroboration / oxidation
Synthesis of cage compounds

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 1 - Repr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-31.0 °F - closed cup

Flash Point(C)

-35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of well-defined polyethylene-based 3-miktoarm star copolymers and terpolymers
Zhang Z, et al.
Macromolecules, 49(7), 2630-2638 (2016)
Amine modified electrospun PIM-1 ultrafine fibers for an efficient removal of methyl orange from an aqueous system
Satilmis B and Uyar T
Applied Surface Science, 453, 220-229 (2018)
Enhancement of CO2 affinity in a polymer of intrinsic microporosity by amine modification
Mason CR, et al.
Macromolecules, 47(3), 1021-1029 (2014)
A Burchert et al.
Leukemia, 29(6), 1331-1335 (2015-02-26)
A minority of chronic myeloid leukemia (CML) patients is capable of successfully discontinuing imatinib. Treatment modalities to increase this proportion are currently unknown. Here, we assessed the role of interferon alpha 2a (IFN) on therapy discontinuation in a previously reported
Kathleen C Rollet-Kurhajec et al.
PloS one, 10(6), e0129868-e0129868 (2015-06-20)
In Hepatitis C virus (HCV) mono-infection, male sex is associated with faster liver fibrosis progression but the effects of sex have not been well studied in HIV-HCV co-infected patients. We examined the influence of sex on progression to significant liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service