Skip to Content
MilliporeSigma
All Photos(1)

Documents

446963

Sigma-Aldrich

Trichloro(hexyl)silane

97%

Synonym(s):

Hexyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5SiCl3
CAS Number:
Molecular Weight:
219.61
Beilstein:
1739071
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.443 (lit.)

bp

191-192 °C (lit.)

density

1.107 g/mL at 25 °C (lit.)

SMILES string

CCCCCC[Si](Cl)(Cl)Cl

InChI

1S/C6H13Cl3Si/c1-2-3-4-5-6-10(7,8)9/h2-6H2,1H3

InChI key

LFXJGGDONSCPOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloro(hexyl)silane is a trichloroalkylsilane that can be used as a surfactant and a capping agent that functionalizes the surface atoms and attaches hexyl groups on silica substrates like silica nanoparticles (SiNPs).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of alkyl chain length on the level of capping of silicon nanoparticles produced by a one-pot synthesis route based on the chemical reduction of micelle
Ashby SP, et al.
Journal of Nanoparticle Research, 15(2), 1425-1425 (2013)
Investigation on hole drift mobility in poly (n-hexylphenylsilane)
Kunimi Y, et al.
Solid State Communications, 114(9), 469-472 (2000)
Simple and facile approach to create charge reversible pores via hydrophobic anchoring of ionic amphiphiles
Sonu KP, et al.
ACS Applied Materials & Interfaces, 9(10), 9136-9142 (2017)
Tatsuya Sugihara et al.
Science advances, 6(51) (2020-12-18)
Adsorbed films often influence mechanical behavior of surfaces, leading to well-known mechanochemical phenomena such as liquid metal embrittlement and environment-assisted cracking. Here, we demonstrate a mechanochemical phenomenon wherein adsorbed long-chain organic monolayers disrupt large-strain plastic deformation in metals. Using high-speed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service