Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44696

Sigma-Aldrich

2-(Dimethylaminomethyl)ferrocen-1-yl-palladium(II) chloride Dinorbornylphosphine Complex

≥97.0% (C)

Synonym(s):

Chloro-2-(dimethylaminomethyl)-ferrocen-1-yl-(dinorbornylphosphine)palladium, SK-CC02-A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39ClFeNPPd
CAS Number:
Molecular Weight:
606.30
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (C)

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

~156 °C (dec.)
~156 °C

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.CN(C)C[C]2[CH][CH][CH][C]2[Pd]Cl.[H][C@]34CC[C@]([H])(C3)C(C4)PC5C[C@]6([H])CC[C@@]5([H])C6

InChI

1S/C14H23P.C8H11N.C5H5.ClH.Fe.Pd/c1-3-11-5-9(1)7-13(11)15-14-8-10-2-4-12(14)6-10;1-9(2)7-8-5-3-4-6-8;1-2-4-5-3-1;;;/h9-15H,1-8H2;3-5H,7H2,1-2H3;1-5H;1H;;/q;;;;;+1/p-1/t9-,10+,11+,12-,13?,14?;;;;;

InChI key

WLPBCKROBGAVNZ-HHYUVNRKSA-M

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Catalyst for CN and CC coupling, especially Buchwald-Hartwig Amination; Heck and Suzuki coupling reactions are also possible. The catalyst is stable to air

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Palladacyclic catalysts by Bedford's group facilitate coupling reactions with low catalyst loadings and challenging aryl chlorides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service