Skip to Content
MilliporeSigma
All Photos(1)

Documents

446947

Sigma-Aldrich

1-Propargyl-1H-benzotriazole

96%

Synonym(s):

1-(2-Propynyl)-1H-benzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N3
CAS Number:
Molecular Weight:
157.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

reaction suitability

reaction type: C-C Bond Formation

mp

52-56 °C (lit.)

SMILES string

C#CCn1nnc2ccccc12

InChI

1S/C9H7N3/c1-2-7-12-9-6-4-3-5-8(9)10-11-12/h1,3-6H,7H2

InChI key

HGIJKQXOOIWWGO-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of clarithromycin analogs via click chemistry as antibacterial agents
  • Sonogashira coupling
  • Bronsted superacid-catalyzed reactions
  • Hydroboration and hydrostannylation reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service