Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

446807

Sigma-Aldrich

Perfluoropentadecane

≥98%

Synonym(s):

Dotriacontafluoropentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)13CF3
CAS Number:
Molecular Weight:
788.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

bp

228 °C (lit.)

mp

110-113 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C15F32/c16-1(17,2(18,19)4(22,23)6(26,27)8(30,31)10(34,35)12(38,39)14(42,43)44)3(20,21)5(24,25)7(28,29)9(32,33)11(36,37)13(40,41)15(45,46)47

InChI key

QKYNDPROJKKCEU-UHFFFAOYSA-N

General description

Perfluoropentadecane is an alkyl fluorinated building block.

Application

Perfluoropentadecane may be used to compose the FC-70, an inert, high-density fluorinated liquid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey R Millman et al.
Nature materials, 4(1), 98-102 (2004-12-21)
The miniaturization of chemical and biological processes in microfluidic devices and bioarrays is a major technological achievement. Microchips performing multiphase material synthesis operations could be a future step in this trend of miniaturizing technology. Here we show how electrically controlled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service