Skip to Content
MilliporeSigma
All Photos(1)

Documents

446645

Sigma-Aldrich

Cobalt(II) 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine

Dye content 95 %

Synonym(s):

(Hexadecafluorophthalocyaninato)cobalt, Cobalt hexadecafluorophthalocyanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32CoF16N8
CAS Number:
Molecular Weight:
859.30
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Dye content, 95%

mp

>300 °C (lit.)

λmax

678 nm

SMILES string

Fc1c(F)c(F)c2c3nc(nc4n5[Co]n6c(n3)c7c(F)c(F)c(F)c(F)c7c6nc8nc(nc5c9c(F)c(F)c(F)c(F)c49)c%10c(F)c(F)c(F)c(F)c8%10)c2c1F

InChI

1S/C32F16N8.Co/c33-9-1-2(10(34)18(42)17(9)41)26-49-25(1)53-27-3-4(12(36)20(44)19(43)11(3)35)29(50-27)55-31-7-8(16(40)24(48)23(47)15(7)39)32(52-31)56-30-6-5(28(51-30)54-26)13(37)21(45)22(46)14(6)38;/q-2;+2

InChI key

OFILAZNRYPZFNX-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service