Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

446254

Sigma-Aldrich

3-Ethyl-2-methyl-2-(3-methylbutyl)oxazolidine

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23NO
CAS Number:
Molecular Weight:
185.31
EC Number:
MDL number:
UNSPSC Code:
12162002

Assay

90%

impurities

<5% 5-methyl-2-hexanone and 2-(ethylamino)ethanol

refractive index

n20/D 1.442 (lit.)

viscosity

<100 cP, Brookfield(lit.)

bp

209 °C (lit.)

solubility

polyols and organic solvents: soluble

density

0.872 g/mL at 25 °C (lit.)

SMILES string

CCN1CCOC1(C)CCC(C)C

Looking for similar products? Visit Product Comparison Guide

Application

Moisture scavenger.
Reactive diluent for urethane prepolymers. Replaces polyol component.

Features and Benefits

Reaction with water yields a volatile ketone and an amino alcohol.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service