All Photos(1)
About This Item
Linear Formula:
(CH3C6H4N=N)2NCH3
CAS Number:
Molecular Weight:
267.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
mp
138-142 °C (lit.)
functional group
amine
SMILES string
CN(\N=N\c1ccc(C)cc1)\N=N\c2ccc(C)cc2
InChI
1S/C15H17N5/c1-12-4-8-14(9-5-12)16-18-20(3)19-17-15-10-6-13(2)7-11-15/h4-11H,1-3H3/b18-16+,19-17+
InChI key
QYJYKVPNNJFJGE-YWNVXTCZSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
1-Methyl-3-p-Tolyltriazene and its Use in the Esterification of Acids.
White EH, et al.
Organic Syntheses, 102-102 (1968)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service