Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

445673

Sigma-Aldrich

N-Fmoc-L-valinol

97%

Synonym(s):

(S)-2-(Fmoc-amino)-3-methyl-1-butanol, N-(9-Fluorenylmethoxycarbonyl)-L-valinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23NO3
CAS Number:
Molecular Weight:
325.40
Beilstein:
7041232
MDL number:
UNSPSC Code:
12352103

Assay

97%

optical activity

[α]20/D −20°, c = 1 in chloroform

mp

129-133 °C (lit.)

SMILES string

CC(C)[C@@H](CO)NC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C20H23NO3/c1-13(2)19(11-22)21-20(23)24-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19,22H,11-12H2,1-2H3,(H,21,23)/t19-/m1/s1

InChI key

MYMGENAMKAPEMT-LJQANCHMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for the synthesis of enantiopure homo-β-amino acids. Used in the efficient synthesis of enantiopure tetrahydroisoquinolines. Intermediate in the one-pot conversion of amino acid carbamates to N-derivatized 2-oxazolidinones.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huwe, C.M. Blechert, S.
Tetrahedron Letters, 35, 9533-9533 (1994)
Caputo, R. et al.
Tetrahedron, 51, 12337-12337 (1995)
R. Caputo et al.
Tetrahedron Letters, 36, 167-167 (1995)
Tietze, L.F. Burkhardt, O.
Synthesis, 1331-1331 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service