Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

445452

Sigma-Aldrich

1-Hydroxy-7-azabenzotriazole

98%

Synonym(s):

3H-[1,2,3]-Triazolo[4,5-b]pyridin-3-ol, HOAt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
Beilstein:
1211115
MDL number:
UNSPSC Code:
12352200

Assay

98%

mp

213-216 °C (dec.) (lit.)

SMILES string

On1nnc2cccnc12

Legal Information

Sold under agreement with PE Biosystems

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service