Skip to Content
MilliporeSigma
All Photos(1)

Documents

445037

Sigma-Aldrich

O-Ethyl methylphosphonothioate

90%

Synonym(s):

EMPSH, EMPTA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3P(S)(OH)OC2H5
CAS Number:
Molecular Weight:
140.14
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.487 (lit.)

bp

73 °C/1 mmHg (lit.)

density

1.18 g/mL at 25 °C (lit.)

SMILES string

CCOP(C)(S)=O

InChI

1S/C3H9O2PS/c1-3-5-6(2,4)7/h3H2,1-2H3,(H,4,7)

InChI key

XXNRHOAJIUSMOQ-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service