Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444677

Sigma-Aldrich

Poly(vinyl acetate-co-crotonic acid)

beads

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[CH2CH(OCCH3)]9[CH(CH3)CH(CO2H)]1]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

beads

composition

crotonic acid, 10 mol %

acid number

62‑70 mg KOH/g

transition temp

Tg (DSC) 61 °C (onset)

solubility

H2O: soluble (as salt form)
absolute ethanol: soluble

density

1.17 g/mL at 25 °C (lit.)

SMILES string

C\C=C\C(O)=O.CC(=O)OC=C

Application

Fixative and film former in hair care products.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service