Skip to Content
MilliporeSigma
All Photos(2)

Documents

444065

Sigma-Aldrich

Isobutyl(trimethoxy)silane

97%

Synonym(s):

Isobutyltrimethoxylsilane, Isobutyltrimethoxysilane, Trimethoxy(2-methylpropyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2Si(OCH3)3
CAS Number:
Molecular Weight:
178.30
Beilstein:
2233976
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

impurities

<1% methyl alcohol

refractive index

n20/D 1.396 (lit.)

bp

154 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CC(C)C)(OC)OC

InChI

1S/C7H18O3Si/c1-7(2)6-11(8-3,9-4)10-5/h7H,6H2,1-5H3

InChI key

XYJRNCYWTVGEEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isobutyl(trimethoxy)silane-modified substrates were used for the preparation of gold films by absorption and adsorption.{87} It may be used as silane precursors for the synthesis of xerogels.{88}

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Functional Layer-By-Layer Design of Xerogel-Based First-Generation Amperometric Glucose Biosensors.
Poulos NG,e t al.
Langmuir, 31(4), 1547-1555 (2015)
Nonaqueous gold colloids. Investigations of deposition and film growth on organically modified substrates and trapping of molecular gold clusters with an alkyl amine.
Klabunde KJ, et al.
New. J. Chem., 22(11), 1275-1283 (1998)
Benjamin J Privett et al.
Biofouling, 26(8), 973-983 (2010-11-18)
This report details the efficacy of nitric oxide (NO)-releasing xerogel surfaces composed of N-(6-aminohexyl)aminopropyl trimethoxysilane (AHAP3) and isobutyltrimethoxysilane (BTMOS) against Candida albicans adhesion, viability, and biofilm formation. A parallel plate flow cell assay was used to examine the effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service