Skip to Content
MilliporeSigma
All Photos(1)

Documents

444022

Sigma-Aldrich

4′-Aminobenzo-18-crown-6

97%

Synonym(s):

(Benzo-18-crown-6)-4′-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H25NO6
CAS Number:
Molecular Weight:
327.37
Beilstein:
5296463
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

SMILES string

Nc1ccc2OCCOCCOCCOCCOCCOc2c1

InChI

1S/C16H25NO6/c17-14-1-2-15-16(13-14)23-12-10-21-8-6-19-4-3-18-5-7-20-9-11-22-15/h1-2,13H,3-12,17H2

InChI key

PZXYILUXRGTFGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4′-Aminobenzo-18-crown-6 can be used:
  • In the surface modification by coating with dithiocarbamate on AuNPs for use as a calorimetric probe for the detection of Pb(II) ions in wastewater.
  • As a precursor for the preparation of thiophene analogs containing copolymers of crown ether to study the chromic properties.
  • To functionalize the gold surface to fabricate an electrochemical sensor for the specific detection of potassium ions.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of novel polythiophenes bearing oligo (ethylene glycol) spacers and crown ethers
Almeida S, et al.
Synthetic Metals, 159(12), 1215-1223 (2009)
A potassium-selective electrochemical sensor based on crown-ether functionalized self assembled monolayer
Kumbhat S and Singh U
Journal of Electroanalytical Chemistry, 809(12), 31-35 (2018)
Selective visual detection of Pb (II) ion via gold nanoparticles coated with a dithiocarbamate-modified 4?-aminobenzo-18-crown-6
Mehta VN, et al.
Microchimica Acta, 181(15-16), 1905-1915 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service