Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

443972

Sigma-Aldrich

1-Chloro-2,5-difluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3F2
CAS Number:
Molecular Weight:
148.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.478 (lit.)

bp

126-128 °C (lit.)

density

1.352 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(F)c(Cl)c1

Application

Used for the synthesis of fluorocyclohexadienes by electrochemical fluorination.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Appl. Electrochem., 25, 651-651 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service