Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

443824

Sigma-Aldrich

4-Chloro-2,5-difluorobenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(F)2CO2H
CAS Number:
Molecular Weight:
192.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

154-157 °C (lit.)

functional group

carboxylic acid
chloro
fluoro

SMILES string

OC(=O)c1cc(F)c(Cl)cc1F

InChI

1S/C7H3ClF2O2/c8-4-2-5(9)3(7(11)12)1-6(4)10/h1-2H,(H,11,12)

InChI key

PEPCYJSDHYMIFN-UHFFFAOYSA-N

General description

4-Chloro-2,5-difluorobenzoic acid reacts with substituted 2-hydroxy acetophenones in the presence of POCl3 to afford 2-acetylphenyl-4-chloro-2,5-difluorobenzoate.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TiO2/modified natural clay semiconductor as a potential electrode for natural dye-sensitized solar cell.
Saelim N, et al.
Ceramics International, 37(2), 659-663 (2011)
Synthesis and antimicrobial evaluations of some novel fluorinated chromones and pyrazoles.
Gadhaveb A, et al.
Indian Journal of Chemistry, 54, 383-390 (2015)
Synthesis of 4-Chloro-2, 5-difluorobenzoic Acid.
Hao-yu Z.
Chemical World, 1, 010-010 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service