Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442445

Sigma-Aldrich

Triphenylborane solution

0.25 M in THF

Synonym(s):

Triphenylborane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3B
CAS Number:
Molecular Weight:
242.12
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

liquid

concentration

0.25 M in THF

bp

65-67 °C

density

0.898 g/mL at 25 °C

SMILES string

c1ccc(cc1)B(c2ccccc2)c3ccccc3

InChI

1S/C18H15B/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

MXSVLWZRHLXFKH-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service