Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442003

Sigma-Aldrich

trans-2-Butenal diethyl acetal

technical, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH(OC2H5)2
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

90%

refractive index

n20/D 1.412 (lit.)

bp

146-150 °C (lit.)

density

0.831 g/mL at 25 °C (lit.)

SMILES string

[H]\C(C)=C(\[H])C(OCC)OCC

InChI

1S/C8H16O2/c1-4-7-8(9-5-2)10-6-3/h4,7-8H,5-6H2,1-3H3/b7-4+

InChI key

ZUMISMXLQDKQDS-QPJJXVBHSA-N

General description

trans-2-Butenal diethyl acetal is an acetal. Acetals are usually derived from the aldehydes. They have two single-bonded oxygen atoms attached to the same carbon atom. They are stable under basic conditions and they can be hydrolyzed back to the carbonyl compound. Acetals serves as a carbonyl protecting group and also are present as functional groups in glycosidic bonds. trans-2-Butenal diethyl acetal has been synthesized from trans-2-butenal with ethanol in the presence of methanesulfonic acid or methylsulfonate as catalysts.[1]

Application

trans-2-Butenal diethyl acetal may be used in the preparation of 2S,3S-epoxybutan-1-ol, an intermediate in the synthesis of erythromycin antibiotic.[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SYNTHESIS OF trans-2-BUTENAL DIETHYL ACETAL CATALYZED BY METHYLSUFONIC ACID OR METHYLSULFONATE [J].
Lifang Z, et al.
Speciality Petrochemicals, 3, 008-008 (2006)
An Efficient Method for One-Pot Reductive Cleavage of Acetals to Primary Alcohols Using a Bimetal Redox Couple CoCl2. 6H2O-Zn.
Sarma K and Goswami A.
Letters in Organic Chemistry, 6(7), 568-572 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service