Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

441724

Sigma-Aldrich

1,3,4,6-Tetrakis(butoxymethyl)glycoluril

average Mn 550, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H46N4O6
CAS Number:
Molecular Weight:
486.65
EC Number:
MDL number:
UNSPSC Code:
12162002

grade

technical grade

mol wt

average Mn 550

refractive index

n20/D 1.4788 (lit.)

Gardner-Holdt

22.7-36.2 stokes(25 °C)(lit.)

bp

>204 °C (lit.)

solubility

polar organic solvents: soluble

density

1.07 g/mL at 25 °C (lit.)

Application

Cross-linking agent.
Cross-linking agent for water- or solvent-based alkyd, epoxy, epoxy ester, polyester and vinyl polymers used in coatings, enamels and primers.

Features and Benefits

Improves adhesion of cured films, increases corrosion resistance and flexibility. Requires strong acid cross-linking catalyst. Low tendency to self-condense.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service