Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

440663

Sigma-Aldrich

4-(2-Chloroacetamido)benzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CONHC6H4CO2H
CAS Number:
Molecular Weight:
213.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

264 °C (dec.) (lit.)

functional group

amide
carboxylic acid
chloro

SMILES string

OC(=O)c1ccc(NC(=O)CCl)cc1

InChI

1S/C9H8ClNO3/c10-5-8(12)11-7-3-1-6(2-4-7)9(13)14/h1-4H,5H2,(H,11,12)(H,13,14)

InChI key

FNXAWVZXABMYRM-UHFFFAOYSA-N

General description

Synthesis of 4-(2-Chloroacetamido)benzoic acid has been reported. It is reported as key intermediate in the synthesis of series of Matijing-Su (MTS, N-(N-benzoyl-L-phenylalanyl)-O-acetyl-L-phenylalanol) derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingying Qiu et al.
Bioorganic & medicinal chemistry, 19(18), 5352-5360 (2011-08-30)
A series of Matijing-Su (MTS, N-(N-benzoyl-l-phenylalanyl)-O-acetyl-L-phenylalanol) derivatives were synthesized and evaluated for their anti-hepatitis B virus (HBV) activity in 2.2.15 cells. The IC(50) of compounds 14a (0.71 μM), 13c (2.85 μM), 13b (4.37 μM), etc. and the selective index of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service