Skip to Content
MilliporeSigma
All Photos(2)

Documents

440515

Sigma-Aldrich

(S)-4-tert-Butyl-2-oxazolidinone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
Beilstein:
1099836
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]21/D −16°, c = 6 in ethanol

mp

118-120 °C (lit.)

SMILES string

CC(C)(C)[C@H]1COC(=O)N1

InChI

1S/C7H13NO2/c1-7(2,3)5-4-10-6(9)8-5/h5H,4H2,1-3H3,(H,8,9)/t5-/m1/s1

InChI key

WKUHGFGTMLOSKM-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile chiral auxiliary for asymmetric synthesis. For a recent review, see Aldrichimica Acta .

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ager, D.J., et al.
Aldrichimica Acta, 30, 3-3 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service