Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

440264

Sigma-Aldrich

Silicon tetrachloride

≥98%

Synonym(s):

Dow Corning® product Z-1228, STC, Tetrachlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiCl4
CAS Number:
Molecular Weight:
169.90
EC Number:
MDL number:
UNSPSC Code:
12352300

vapor density

5.86 (vs air)

vapor pressure

420 mmHg ( 37.7 °C)

Assay

≥98%

form

liquid

bp

57.6 °C (lit.)

mp

−70 °C (lit.)

density

1.483 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)Cl

InChI

1S/Cl4Si/c1-5(2,3)4

InChI key

FDNAPBUWERUEDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Repackaged by Aldrich Chemical Company, Inc. Manufactured by Dow Corning Corporation, U.S.A.
®Registered trademark of Dow Corning Corporation, U.S.A.
Dow Corning is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service