Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

439630

Sigma-Aldrich

Thulium(III) bromide

anhydrous, powder, 99.99%

Synonym(s):

Thulium tribromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TmBr3
CAS Number:
Molecular Weight:
408.65
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

anhydrous

Assay

99.99%

form

powder

reaction suitability

reagent type: catalyst
core: thulium

mp

952 °C (lit.)

SMILES string

Br[Tm](Br)Br

InChI

1S/3BrH.Tm/h3*1H;/q;;;+3/p-3

InChI key

HQSWGSFQSCMHFQ-UHFFFAOYSA-K

Application

Used for development of advanced ceramic discharge lamps and metal halide lamps

Reagent for complexation of lanthanide bromides with aluminum bromide

Reactant for preparation of alkali metal thulium bromides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service