Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

439401

Sigma-Aldrich

(Phenylthio)acetyl chloride

97%

Synonym(s):

Thiophenoxyacetyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SCH2COCl
CAS Number:
Molecular Weight:
186.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.5825 (lit.)

density

1.258 g/mL at 25 °C (lit.)

functional group

acyl chloride
thioether

SMILES string

ClC(=O)CSc1ccccc1

InChI

1S/C8H7ClOS/c9-8(10)6-11-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

TWOLQIUHVMTAAE-UHFFFAOYSA-N

General description

(Phenylthio)acetyl chloride is an acid halide. It participates in the Friedel -Crafts acylation of arenes to afford α-sulfenylated acetophenones.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[3, 3]-Sigmatropic rearrangements: short, stereocontrolled syntheses of functionalized vitamin D 3 side-chain units.
Hatcher MA and Posner GH.
Tetrahedron Letters, 43(28), 5009-5012 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service