Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

439339

Sigma-Aldrich

1,4-Cyclohexanedimethanol dibenzoate, mixture of cis and trans

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO2CH2)2C6H10
CAS Number:
Molecular Weight:
352.42
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

SMILES string

O=C(OCC1CCC(CC1)COC(=O)c2ccccc2)c3ccccc3

Application

Flow improvement additive in powder coatings and processing aid in plastics.
Plasticizer for hot melt adhesive systems.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service