Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

438375

Sigma-Aldrich

Tetramethylammonium formate solution

30 wt. % in H2O, ≥99.99% trace metals basis

Synonym(s):

Formic acid tetramethylammonium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(HCOO)
CAS Number:
Molecular Weight:
119.16
Beilstein:
3916177
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

≥99.99% trace metals basis

form

liquid

concentration

30 wt. % in H2O

refractive index

n20/D 1.364

bp

102 °C

density

1.02 g/mL at 25 °C

SMILES string

[H]C([O-])=O.C[N+](C)(C)C

InChI

1S/C4H12N.CH2O2/c1-5(2,3)4;2-1-3/h1-4H3;1H,(H,2,3)/q+1;/p-1

InChI key

WWIYWFVQZQOECA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service