Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

437387

Sigma-Aldrich

Menthyl anthranilate

98%

Synonym(s):

Menthyl o-aminobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25NO2
CAS Number:
Molecular Weight:
275.39
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.542 (lit.)

bp

177-179 °C/3 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

CC(C)C1CCC(C)CC1OC(=O)c2ccccc2N

InChI

1S/C17H25NO2/c1-11(2)13-9-8-12(3)10-16(13)20-17(19)14-6-4-5-7-15(14)18/h4-7,11-13,16H,8-10,18H2,1-3H3

InChI key

SOXAGEOHPCXXIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Menthyl anthranilate is a fluorescent agent and a UV filter that can be used as an alternative to oxybenzone. It is majorly used in sunscreen based products. It can be easily mixed in cosmetic oils and can also be emulsified in aqueous formulations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Photophysical Properties of Menthyl Anthranilate: A UV-A Sunscreen.
Beeby A and Jones AE
Photochemistry and Photobiology, 72(1), 10-15 (2000)
The feasibility of using fluorescence spectroscopy as a rapid, non-invasive method for evaluating sunscreen performance.
Stokes RP and Diffey BL
Journal of Photochemistry and Photobiology. B, Biology, 50(2-3), 137-143 (1999)
Marlene Vila et al.
Journal of chromatography. A, 1405, 12-22 (2015-06-21)
A methodology based on pressurized liquid extraction (PLE) followed by gas chromatography-tandem mass spectrometry (GC-MS/MS) has been developed for the simultaneous analysis of different classes of UV filters including methoxycinnamates, benzophenones, salicylates, p-aminobenzoic acid derivatives, and others in cosmetic products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service