Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

437085

Sigma-Aldrich

5-O-(tert-Butyldimethylsilyl)-2,3-O-isopropylidene-D-ribonic acid γ-lactone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26O5Si
CAS Number:
Molecular Weight:
302.44
Beilstein:
4296295
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D −47°, c = 0.8 in chloroform

mp

72-74 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)OC[C@H]1OC(=O)[C@@H]2OC(C)(C)O[C@H]12

InChI

1S/C14H26O5Si/c1-13(2,3)20(6,7)16-8-9-10-11(12(15)17-9)19-14(4,5)18-10/h9-11H,8H2,1-7H3/t9-,10-,11-/m1/s1

InChI key

VBPYRQYBEKHPKA-GMTAPVOTSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Wang et al.
Molecular cell, 74(4), 742-757 (2019-04-14)
Disturbances in autophagy and stress granule dynamics have been implicated as potential mechanisms underlying inclusion body myopathy (IBM) and related disorders. Yet the roles of core autophagy proteins in IBM and stress granule dynamics remain poorly characterized. Here, we demonstrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service