Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

437042

Supelco

N-Isobutyryl-D-cysteine

97%, ChiraSelect

Synonym(s):

N-(2-Methylpropionyl)-D-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH[NHCOCH(CH3)2]CO2H
CAS Number:
Molecular Weight:
191.25
MDL number:
UNSPSC Code:
12000000
eCl@ss:
32160406
PubChem Substance ID:

product line

ChiraSelect

Assay

97%

mp

97-101 °C (lit.)

SMILES string

CC(C)C(=O)N[C@H](CS)C(O)=O

InChI

1S/C7H13NO3S/c1-4(2)6(9)8-5(3-12)7(10)11/h4-5,12H,3H2,1-2H3,(H,8,9)(H,10,11)/t5-/m1/s1

InChI key

BWBQXMAXLAHHTK-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service