Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

436399

Sigma-Aldrich

Poly(4-vinylpyridine), cross-linked, methyl chloride quaternary salt

Reillex® HPQ ion-exchange resin

Synonym(s):

Reillex® HPQ

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

form

beads

composition

water, 55 wt. %

manufacturer/tradename

Reillex® HPQ ion-exchange resin

particle size

300-1,000 μm

capacity

3.4 meq/g exchange capacity(dry)

solubility

water, acids, bases and organic solvents: insoluble

Looking for similar products? Visit Product Comparison Guide

Application

Anionic ion exchange resin.

Legal Information

Reillex is a registered trademark of Vertellus Specialties, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service