Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

436208

Sigma-Aldrich

Vinyl stearate

contains 20 ppm MEHQ as inhibitor, 95%

Synonym(s):

Vinyl octadecanoate, Vinyl stearate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CO2CH=CH2
CAS Number:
Molecular Weight:
310.51
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

contains

20 ppm MEHQ as inhibitor

bp

385 °C (lit.)

mp

35-37 °C (lit.)

density

0.904 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC=C

InChI

1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h4H,2-3,5-19H2,1H3

InChI key

AFSIMBWBBOJPJG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Eunae Cho et al.
Scientific reports, 6, 23740-23740 (2016-03-30)
Amphiphilic cyclodextrins have been synthesized with self-acylating reaction using vinyl esters in dimethylformamide. In the present study no base, catalyst, or enzyme was used, and the structural analyses using thin layer chromatography, nuclear magnetic resonance spectroscopy and mass spectrometry show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service