Skip to Content
MilliporeSigma
All Photos(2)

Documents

436054

Sigma-Aldrich

Indium(III) fluoride trihydrate

99%

Synonym(s):

Indium trifluoride trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
InF3 · 3H2O
CAS Number:
Molecular Weight:
225.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder and chunks

reaction suitability

reagent type: catalyst
core: indium

solubility

H2O: slightly soluble(lit.)

SMILES string

O.O.O.F[In](F)F

InChI

1S/3FH.In.3H2O/h3*1H;;3*1H2/q;;;+3;;;/p-3

InChI key

FRJQEDQNNQVYRT-UHFFFAOYSA-K

Application

Promotes chemoselective addition of TMSCN to aldehydes forming the respective cyanohydrins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loh T-P. et al.
Synlett, 369-369 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service