Skip to Content
MilliporeSigma
All Photos(1)

Documents

435996

Sigma-Aldrich

Tin(II) tetrafluoroborate solution

50 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sn(BF4)2
CAS Number:
Molecular Weight:
292.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

concentration

50 wt. % in H2O

density

1.67 g/mL at 25 °C

SMILES string

[SnH2++].F[B-](F)(F)F.F[B-](F)(F)F

Application

Employed in metal plating.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prod. Finish., 58, 175-175 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service