Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

434957

Sigma-Aldrich

Poly(acrylamide-co-acrylic acid)

average Mw 15,000,000, granular

Synonym(s):

Acrylamide-acrylic acid copolymer, Acrylamide-acrylic acid resin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

granular

mol wt

average Mw 15,000,000

composition

acrylic acid, 1.5 wt. %

impurities

15 wt. % adsorbed water

pH

5-7

viscosity

4-5 cP, 0.1 % in H2O

solubility

water and some protic solvents: soluble

density

0.75 g/mL at 25 °C (lit.)

SMILES string

NC(=O)C=C.OC(=O)C=C

InChI

1S/C3H5NO.C3H4O2/c2*1-2-3(4)5/h2H,1H2,(H2,4,5);2H,1H2,(H,4,5)

InChI key

RNIHAPSVIGPAFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thickening agent, binder, sizing agent, flocculating agent, crosslinker, filtering aid, lubricant, drag reduction and polymer recovery agent.

Features and Benefits

Changes in pH from 3 to 10 have little effect on viscosity. High tolerance for electrolytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service