Skip to Content
MilliporeSigma
All Photos(1)

Documents

434752

Sigma-Aldrich

Poly(ethylene-co-1-butene-co-1-hexene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)x[CH2CH(C2H5)]y[CH2CH[(CH2)3CH3]]Z
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

gloss

95.5 % , ASTM D 2457)

melt index

3.5 g/10 min (190 °C/2.16kg)

contains

antioxidant (varying amounts)

impact strength

>1450 g/mil (ASTM D 1709, dart drop, F50)

mp

92 °C (lit.)

transition temp

Tm 95 °C (DSC)

density

0.9 g/mL at 25 °C (lit.)

Mw/Mn

~2.0

elongation

linear, ASTM D 882 - 350% (at break)

SMILES string

C=C.CCC=C.CCCCC=C

Looking for similar products? Visit Product Comparison Guide

Application

Mono- or multilayer co-extruded cast films. Packaging seal layers and laminated film.

Physical properties

Linear terpolymer.
Narrower molecular weight and composition distributions than obtained with conventional initiators. Good toughness and heat sealing properties.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service