Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

434299

Sigma-Aldrich

Boron trifluoride phenol complex (1:2)

25-27% BF3 basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · 2C6H5OH
CAS Number:
Molecular Weight:
256.03
UNSPSC Code:
12352106
PubChem Substance ID:

form

liquid

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

concentration

25-27% boron trifluoride

bp

178 °C (lit.)

density

1.27 g/mL at 25 °C (lit.)

SMILES string

FB(F)F.Oc1ccccc1.Oc2ccccc2

InChI

1S/2C6H6O.BF3/c2*7-6-4-2-1-3-5-6;2-1(3)4/h2*1-5,7H;

InChI key

OQORKFSQJPQHSZ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service